Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281710.1 | NP_001268639.1 | 2 - 3 | 330 | 91 |
NM_001281711.1 | NP_001268640.1 | 2 - 3 | 291 | 91 | |
NM_015341.4 | NP_056156.2 | 2 - 3 | 363 | 91 | |
XM_005263908.3 | XP_005263965.1 | 2 - 3 | 303 | 91 | |
XM_006712388.3 | XP_006712451.1 | 2 - 3 | 303 | 91 | |
GenBank mRNA | AK291053.1 | - | 2 - 3 | 304 | 91 |
AK299248.1 | - | 2 - 3 | 232 | 91 | |
AK308050.1 | - | 2 - 3 | 270 | 91 | |
AK310649.1 | - | 2 - 3 | 231 | 91 | |
BC024211.2 | - | 2 - 3 | 317 | 91 | |
BI559873.1 | - | 2 - 3 | 351 | 91 | |
BI560881.1 | - | 2 - 3 | 368 | 91 | |
D38553.1 | - | 2 - 3 | 293 | 91 |