Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281710.1 | NP_001268639.1 | 4 - 5 | 514 | 61 |
NM_001281711.1 | NP_001268640.1 | 4 - 5 | 475 | 61 | |
NM_001281712.1 | NP_001268641.1 | 3 - 4 | 294 | 61 | |
NM_015341.4 | NP_056156.2 | 4 - 5 | 547 | 61 | |
XM_005263908.3 | XP_005263965.1 | 4 - 5 | 487 | 61 | |
XM_006712388.3 | XP_006712451.1 | 4 - 5 | 487 | 61 | |
GenBank mRNA | AK291053.1 | - | 4 - 5 | 488 | 61 |
AK299248.1 | - | 4 - 5 | 416 | 61 | |
AK303725.1 | - | 3 - 4 | 239 | 61 | |
AK308050.1 | - | 4 - 5 | 454 | 61 | |
AK310649.1 | - | 4 - 5 | 415 | 61 | |
BC024211.2 | - | 4 - 5 | 501 | 61 | |
BI559873.1 | - | 4 - 5 | 534 | 61 | |
BI560881.1 | - | 4 - 5 | 552 | 61 | |
D38553.1 | - | 4 - 5 | 477 | 61 |