Assay Design
Gene Transcripts
- Gene Symbol
- COX6B2
- Entrez Gene ID
-
125965
- Gene Name
- cytochrome c oxidase subunit 6B2
- Gene Aliases
-
COXVIB2, CT59
- Chromosome Location
-
Chr.19: 55349702 - 55354814 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK057427.1 |
- |
4 - 5 |
467 |
58 |
View Full Product Details