Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013703.3 | NP_001013725.2 | 37 - 38 | 4844 | 92 |
XM_011521600.2 | XP_011519902.1 | 36 - 37 | 4696 | 92 | |
GenBank mRNA | AB037759.1 | - | 33 - 34 | 4310 | 92 |
AJ243428.1 | - | 15 - 16 | 1471 | 92 | |
AK027011.1 | - | 23 - 24 | 2282 | 92 | |
AK074361.1 | - | 15 - 16 | 1512 | 92 | |
AL137627.1 | - | 6 - 7 | 398 | 92 | |
AL157497.1 | - | 26 - 27 | 2641 | 92 | |
AL832907.1 | - | 31 - 32 | 4104 | 92 | |
BC009350.2 | - | 23 - 24 | 2314 | 92 | |
BX649048.1 | - | 20 - 21 | 3689 | 92 |