Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017425.3 | NP_059121.1 | 3 - 4 | 364 | 131 |
XM_011542870.2 | XP_011541172.1 | 3 - 4 | 554 | 131 | |
GenBank mRNA | AF334735.1 | - | 3 - 4 | 363 | 131 |
AK311806.1 | - | 3 - 4 | 307 | 131 | |
BC019224.1 | - | 3 - 4 | 296 | 131 | |
BC032457.2 | - | 2 - 3 | 256 | 131 | |
DA511916.1 | - | 3 - 4 | 413 | 131 | |
KU257683.1 | - | 2 - 3 | 228 | 131 |