Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018944.2 | NP_061817.1 | 4 - 5 | 673 | 129 |
GenBank mRNA | AF231921.1 | - | 4 - 5 | 645 | 129 |
AF387845.1 | - | 4 - 5 | 643 | 129 | |
AK075281.1 | - | 4 - 5 | 645 | 129 | |
AK312073.1 | - | 4 - 5 | 764 | 129 | |
AW962893.1 | - | 2 - 3 | 127 | 129 | |
BC042917.1 | - | 4 - 5 | 649 | 129 | |
BU567844.1 | - | 2 - 3 | 240 | 129 |