Assay Design



Gene Transcripts

Gene Symbol
RAD23B
Entrez Gene ID
5887
Gene Name
RAD23 homolog B, nucleotide excision repair protein
Gene Aliases
HHR23B, HR23B, P58
Chromosome Location
Chr.9: 107283236 - 107332194 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244713.1 NP_001231642.1 6 - 7 764 65
NM_001244724.1 NP_001231653.1 6 - 7 754 65
NM_002874.4 NP_002865.1 6 - 7 1079 65
GenBank mRNA AK125226.1 - 7 - 8 983 65
AK293532.1 - 6 - 7 1071 65
AK297986.1 - 6 - 7 724 65
AK316189.1 - 6 - 7 679 65
AY313777.1 - 6 - 7 754 65
BC020973.2 - 6 - 7 735 65
D21090.1 - 6 - 7 998 65

View Full Product Details