Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002705.4 | NP_002696.3 | 21 - 22 | 2695 | 71 |
XM_006720902.3 | XP_006720965.1 | 21 - 22 | 2659 | 71 | |
XM_017023374.1 | XP_016878863.1 | 21 - 22 | 2707 | 71 | |
XM_017023375.1 | XP_016878864.1 | 21 - 22 | 2719 | 71 | |
GenBank mRNA | AB011140.1 | - | 12 - 13 | 1617 | 71 |
AF001691.1 | - | 21 - 22 | 2695 | 71 | |
AF013717.1 | - | 21 - 22 | 2616 | 71 | |
AK295623.1 | - | 9 - 10 | 1136 | 71 | |
BC114620.1 | - | 21 - 22 | 2655 | 71 |