Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002705.4 | NP_002696.3 | 3 - 4 | 414 | 77 |
XM_006720902.3 | XP_006720965.1 | 3 - 4 | 378 | 77 | |
XM_017023374.1 | XP_016878863.1 | 3 - 4 | 378 | 77 | |
XM_017023375.1 | XP_016878864.1 | 3 - 4 | 390 | 77 | |
GenBank mRNA | AF001691.1 | - | 3 - 4 | 414 | 77 |
AF013717.1 | - | 3 - 4 | 335 | 77 | |
AK295623.1 | - | 3 - 4 | 440 | 77 | |
BC114620.1 | - | 3 - 4 | 374 | 77 |