Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005188.3 | NP_005179.2 | 2 - 3 | 588 | 96 |
XM_011543057.2 | XP_011541359.1 | 2 - 3 | 588 | 96 | |
GenBank mRNA | BC132733.1 | - | 2 - 3 | 517 | 96 |
BC136463.1 | - | 2 - 3 | 517 | 96 | |
X57110.1 | - | 2 - 3 | 594 | 96 |