Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005188.3 | NP_005179.2 | 9 - 10 | 1570 | 137 |
XM_017018511.1 | XP_016874000.1 | 8 - 9 | 1005 | 137 | |
GenBank mRNA | BC132733.1 | - | 9 - 10 | 1499 | 137 |
BC136463.1 | - | 9 - 10 | 1499 | 137 | |
X57110.1 | - | 9 - 10 | 1576 | 137 |