Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005214.4 | NP_005205.2 | 2 - 3 | 617 | 70 |
GenBank mRNA | AF414120.1 | - | 2 - 3 | 617 | 70 |
AK313732.1 | - | 2 - 3 | 602 | 70 | |
AY209009.1 | - | 2 - 3 | 462 | 70 | |
BC069566.1 | - | 2 - 3 | 557 | 70 | |
BC070162.1 | - | 2 - 3 | 618 | 70 | |
BC074842.2 | - | 2 - 3 | 488 | 70 | |
BC074893.2 | - | 2 - 3 | 493 | 70 | |
L15006.1 | - | 2 - 3 | 462 | 70 |