Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000444.5 | NP_000435.3 | 9 - 10 | 1640 | 159 |
NM_001282754.1 | NP_001269683.1 | 9 - 10 | 1640 | 159 | |
XM_011545533.1 | XP_011543835.1 | 5 - 6 | 449 | 159 | |
XM_017029579.1 | XP_016885068.1 | 5 - 6 | 450 | 159 | |
GenBank mRNA | AD000712.1 | - | 9 - 10 | 1237 | 159 |
AK298030.1 | - | 9 - 10 | 1186 | 159 | |
AK301528.1 | - | 8 - 9 | 1376 | 159 | |
AK304551.1 | - | 2 - 3 | 477 | 159 | |
BC105057.1 | - | 9 - 10 | 1162 | 159 | |
BC105059.1 | - | 9 - 10 | 1162 | 159 | |
U75645.1 | - | 9 - 10 | 1154 | 159 | |
U82970.1 | - | 9 - 10 | 1681 | 159 | |
U87284.1 | - | 9 - 10 | 1279 | 159 |