Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173854.5 | NP_776253.3 | 5 - 6 | 1736 | 69 |
XM_005245069.2 | XP_005245126.1 | 5 - 6 | 1412 | 69 | |
GenBank mRNA | AJ514402.1 | - | 4 - 5 | 823 | 69 |
AK125075.1 | - | 4 - 5 | 497 | 69 | |
AK128069.1 | - | 4 - 5 | 989 | 69 | |
AK314732.1 | - | 4 - 5 | 894 | 69 | |
AL831974.1 | - | 5 - 6 | 1572 | 69 | |
AL832001.1 | - | 5 - 6 | 1572 | 69 | |
AL832362.1 | - | 5 - 6 | 1572 | 69 | |
BC136303.1 | - | 4 - 5 | 1217 | 69 | |
BX641054.1 | - | 5 - 6 | 1574 | 69 | |
BX648979.1 | - | 5 - 6 | 1610 | 69 |