Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282622.1 | NP_001269551.1 | 24 - 25 | 4651 | 72 |
NM_004187.3 | NP_004178.2 | 24 - 25 | 4654 | 72 | |
XM_011530824.2 | XP_011529126.1 | 24 - 25 | 4389 | 72 | |
XM_011530825.2 | XP_011529127.1 | 23 - 24 | 4344 | 72 | |
XM_011530827.2 | XP_011529129.1 | 24 - 25 | 4467 | 72 | |
XM_011530828.1 | XP_011529130.1 | 24 - 25 | 4467 | 72 | |
XM_011530829.2 | XP_011529131.1 | 25 - 26 | 4536 | 72 | |
XM_011530830.2 | XP_011529132.1 | 25 - 26 | 4483 | 72 | |
XM_011530831.2 | XP_011529133.1 | 18 - 19 | 3475 | 72 | |
XM_017029886.1 | XP_016885375.1 | 24 - 25 | 4464 | 72 | |
GenBank mRNA | BC054499.2 | - | 24 - 25 | 4405 | 72 |
EF613277.1 | - | 24 - 25 | 4121 | 72 | |
L25270.1 | - | 24 - 25 | 4652 | 72 |