Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000466.2 | NP_000457.1 | 21 - 22 | 3534 | 88 |
NM_001282677.1 | NP_001269606.1 | 20 - 21 | 3363 | 88 | |
NM_001282678.1 | NP_001269607.1 | 21 - 22 | 3569 | 88 | |
XM_017012319.1 | XP_016867808.1 | 16 - 17 | 2454 | 88 | |
GenBank mRNA | AB008112.1 | - | 21 - 22 | 3505 | 88 |
AB052090.1 | - | 20 - 21 | 3267 | 88 | |
AB052091.1 | - | 21 - 22 | 3438 | 88 | |
AB052092.1 | - | 21 - 22 | 3438 | 88 | |
AB052093.1 | - | 21 - 22 | 3438 | 88 | |
AB052094.1 | - | 21 - 22 | 3438 | 88 | |
AB209355.1 | - | 9 - 10 | 4493 | 88 | |
AF026086.1 | - | 21 - 22 | 3498 | 88 | |
AF030356.1 | - | 21 - 22 | 3442 | 88 | |
AK292955.1 | - | 21 - 22 | 3439 | 88 | |
AK293757.1 | - | 21 - 22 | 3477 | 88 | |
AK295686.1 | - | 18 - 19 | 2478 | 88 | |
BC035575.1 | - | 21 - 22 | 3476 | 88 |