Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000466.2 | NP_000457.1 | 6 - 7 | 1456 | 127 |
NM_001282677.1 | NP_001269606.1 | 6 - 7 | 1456 | 127 | |
NM_001282678.1 | NP_001269607.1 | 6 - 7 | 1491 | 127 | |
XM_017012319.1 | XP_016867808.1 | 2 - 3 | 459 | 127 | |
GenBank mRNA | AB008112.1 | - | 6 - 7 | 1427 | 127 |
AB052090.1 | - | 6 - 7 | 1360 | 127 | |
AB052091.1 | - | 6 - 7 | 1360 | 127 | |
AB052092.1 | - | 6 - 7 | 1360 | 127 | |
AB052093.1 | - | 6 - 7 | 1360 | 127 | |
AB052094.1 | - | 6 - 7 | 1360 | 127 | |
AF026086.1 | - | 6 - 7 | 1420 | 127 | |
AF030356.1 | - | 6 - 7 | 1364 | 127 | |
AK292955.1 | - | 6 - 7 | 1361 | 127 | |
AK293757.1 | - | 6 - 7 | 1399 | 127 | |
AK293848.1 | - | 2 - 3 | 261 | 127 | |
AK295686.1 | - | 3 - 4 | 400 | 127 | |
BC035575.1 | - | 6 - 7 | 1398 | 127 |