Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098576.1 | NP_001092046.1 | 8 - 9 | 1117 | 104 |
NM_003217.2 | NP_003208.2 | 8 - 9 | 747 | 104 | |
XM_005269126.3 | XP_005269183.1 | 9 - 10 | 1019 | 104 | |
GenBank mRNA | AF033095.1 | - | 8 - 9 | 677 | 104 |
AK124849.1 | - | 8 - 9 | 1442 | 104 | |
AK130050.1 | - | 8 - 9 | 700 | 104 | |
AK130462.1 | - | 6 - 7 | 569 | 104 | |
AK225804.1 | - | 8 - 9 | 700 | 104 | |
AK299587.1 | - | 7 - 8 | 582 | 104 | |
AK312238.1 | - | 8 - 9 | 683 | 104 | |
AY736129.1 | - | 7 - 8 | 615 | 104 | |
BC000916.1 | - | 8 - 9 | 680 | 104 | |
BC036203.1 | - | 8 - 9 | 755 | 104 | |
X75861.1 | - | 7 - 8 | 655 | 104 |