Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014957.2 | NP_055772.2 | 16 - 17 | 2874 | 84 |
XM_005250838.4 | XP_005250895.2 | 16 - 17 | 2765 | 84 | |
XM_005250839.2 | XP_005250896.2 | 16 - 17 | 2765 | 84 | |
XM_005250840.4 | XP_005250897.2 | 15 - 16 | 2609 | 84 | |
XM_005250841.3 | XP_005250898.2 | 15 - 16 | 2609 | 84 | |
XM_005250842.4 | XP_005250899.1 | 14 - 15 | 2626 | 84 | |
XM_011516933.1 | XP_011515235.1 | 16 - 17 | 2765 | 84 | |
XM_011516934.2 | XP_011515236.1 | 16 - 17 | 2765 | 84 | |
XM_011516937.1 | XP_011515239.1 | 16 - 17 | 2765 | 84 | |
XM_011516938.2 | XP_011515240.1 | 9 - 10 | 1920 | 84 | |
XM_011516939.2 | XP_011515241.1 | 7 - 8 | 1443 | 84 | |
XM_011516940.2 | XP_011515242.1 | 7 - 8 | 1420 | 84 | |
XM_017013241.1 | XP_016868730.1 | 16 - 17 | 2767 | 84 | |
XM_017013242.1 | XP_016868731.1 | 17 - 18 | 3128 | 84 | |
XM_017013243.1 | XP_016868732.1 | 8 - 9 | 1880 | 84 | |
GenBank mRNA | AB020677.2 | - | 11 - 12 | 2061 | 84 |
BC133023.1 | - | 14 - 15 | 2461 | 84 | |
BC144226.1 | - | 14 - 15 | 2461 | 84 |