Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033402.4 | NP_208325.3 | 10 - 11 | 1829 | 129 |
XM_005251318.3 | XP_005251375.1 | 9 - 10 | 1618 | 129 | |
XM_005251319.4 | XP_005251376.1 | 8 - 9 | 1312 | 129 | |
XM_005251320.3 | XP_005251377.1 | 7 - 8 | 1104 | 129 | |
XM_005251322.3 | XP_005251379.1 | 6 - 7 | 918 | 129 | |
XM_017013920.1 | XP_016869409.1 | 11 - 12 | 1777 | 129 | |
XM_017013921.1 | XP_016869410.1 | 11 - 12 | 1777 | 129 | |
XM_017013922.1 | XP_016869411.1 | 11 - 12 | 1777 | 129 | |
XM_017013923.1 | XP_016869412.1 | 10 - 11 | 1604 | 129 | |
XM_017013924.1 | XP_016869413.1 | 11 - 12 | 1777 | 129 | |
XM_017013925.1 | XP_016869414.1 | 8 - 9 | 1323 | 129 | |
XM_017013926.1 | XP_016869415.1 | 8 - 9 | 1306 | 129 | |
XM_017013927.1 | XP_016869416.1 | 9 - 10 | 1334 | 129 | |
XM_017013928.1 | XP_016869417.1 | 6 - 7 | 786 | 129 | |
GenBank mRNA | AB051551.1 | - | 9 - 10 | 2450 | 129 |
AK056185.1 | - | 9 - 10 | 129 | ||
AK300874.1 | - | 9 - 10 | 1473 | 129 | |
AK302648.1 | - | 10 - 11 | 1679 | 129 | |
AK307443.1 | - | 7 - 8 | 1085 | 129 | |
AK307636.1 | - | 9 - 10 | 1463 | 129 | |
BC030701.2 | - | 10 - 11 | 1702 | 129 |