Assay Design



Gene Transcripts

Gene Symbol
GALC
Entrez Gene ID
2581
Gene Name
galactosylceramidase
Gene Aliases
-
Chromosome Location
Chr.14: 87933014 - 87993665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000153.3 NP_000144.2 4 - 5 554 82
NM_001201401.1 NP_001188330.1 3 - 4 485 82
NM_001201402.1 NP_001188331.1 4 - 5 535 82
XM_011536618.2 XP_011534920.1 4 - 5 836 82
GenBank mRNA AK294040.1 - 4 - 5 461 82
AK296530.1 - 3 - 4 376 82
AK302683.1 - 4 - 5 708 82
AK302956.1 - 4 - 5 374 82
BC036518.2 - 4 - 5 406 82
D25283.1 - 4 - 5 437 82
D25284.1 - 4 - 5 437 82
DA247447.1 - 4 - 5 478 82
DA332567.1 - 4 - 5 518 82
DC334180.1 - 4 - 5 535 82
L23116.1 - 4 - 5 446 82

View Full Product Details