Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022133.3 | NP_071416.2 | 3 - 4 | 875 | 159 |
NM_152836.2 | NP_690049.1 | 2 - 3 | 629 | 159 | |
XM_005251282.4 | XP_005251339.1 | 3 - 4 | 582 | 159 | |
XM_005251283.2 | XP_005251340.1 | 4 - 5 | 719 | 159 | |
GenBank mRNA | AK290903.1 | - | 2 - 3 | 545 | 159 |
BC033630.1 | - | 2 - 3 | 537 | 159 | |
CB962009.1 | - | 2 - 3 | 607 | 159 |