Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015346.3 | NP_056161.2 | 39 - 40 | 7327 | 91 |
XM_017021124.1 | XP_016876613.1 | 39 - 40 | 7346 | 91 | |
XM_017021125.1 | XP_016876614.1 | 39 - 40 | 7346 | 91 | |
XM_017021126.1 | XP_016876615.1 | 38 - 39 | 7960 | 91 | |
XM_017021127.1 | XP_016876616.1 | 28 - 29 | 5042 | 91 | |
XM_017021128.1 | XP_016876617.1 | 28 - 29 | 4895 | 91 | |
GenBank mRNA | AB002319.1 | - | 24 - 25 | 4197 | 91 |
AB425197.1 | - | 39 - 40 | 7315 | 91 | |
AK024743.1 | - | 11 - 12 | 1689 | 91 | |
BC008927.2 | - | 5 - 6 | 726 | 91 | |
BC033235.1 | - | 6 - 7 | 1022 | 91 | |
BC144594.1 | - | 37 - 38 | 6631 | 91 | |
BX537886.1 | - | 39 - 40 | 7321 | 91 | |
BX538025.1 | - | 35 - 36 | 6820 | 91 | |
CR749276.1 | - | 40 - 41 | 7262 | 91 | |
CR749365.1 | - | 38 - 39 | 7940 | 91 |