Assay Design



Gene Transcripts

Gene Symbol
RMND1
Entrez Gene ID
55005
Gene Name
required for meiotic nuclear division 1 homolog
Gene Aliases
C6orf96, COXPD11, RMD1, bA351K16, bA351K16.3
Chromosome Location
Chr.6: 151404548 - 151452181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271937.1 NP_001258866.1 9 - 10 860 103
NM_017909.3 NP_060379.2 10 - 11 1378 103
XM_005267040.3 XP_005267097.1 9 - 10 859 103
XM_017010988.1 XP_016866477.1 10 - 11 1377 103
GenBank mRNA AK000634.1 - 10 - 11 1321 103
AK026480.1 - 9 - 10 879 103
AK292339.1 - 10 - 11 1353 103
BC106065.1 - 10 - 11 1340 103
BC119683.1 - 10 - 11 1243 103

View Full Product Details