Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127713.1 | NP_001121185.1 | 2 - 3 | 453 | 87 |
NM_015915.4 | NP_056999.2 | 1 - 2 | 269 | 87 | |
NM_181598.3 | NP_853629.2 | 1 - 2 | 269 | 87 | |
GenBank mRNA | AF444143.1 | - | 1 - 2 | 200 | 87 |
AK223436.1 | - | 1 - 2 | 182 | 87 | |
AK290185.1 | - | 1 - 2 | 182 | 87 | |
AK312518.1 | - | 1 - 2 | 158 | 87 | |
AL833591.1 | - | 2 - 3 | 237 | 87 | |
AY032844.1 | - | 1 - 2 | 197 | 87 | |
BC010708.2 | - | 1 - 2 | 172 | 87 | |
BG704589.1 | - | 1 - 2 | 257 | 87 | |
BI601372.1 | - | 1 - 2 | 271 | 87 | |
DA088230.1 | - | 1 - 2 | 269 | 87 | |
DA397302.1 | - | 2 - 3 | 509 | 87 |