Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308020.1 | NP_001294949.1 | 13 - 14 | 1492 | 87 |
NM_002647.3 | NP_002638.2 | 14 - 15 | 1681 | 87 | |
XM_011526031.2 | XP_011524333.1 | 15 - 16 | 1846 | 87 | |
GenBank mRNA | AK090567.1 | - | 1 - 2 | 2215 | 87 |
AK298517.1 | - | 13 - 14 | 1436 | 87 | |
BC033004.2 | - | 14 - 15 | 1631 | 87 | |
BC053651.1 | - | 14 - 15 | 1611 | 87 | |
Z46973.1 | - | 14 - 15 | 1634 | 87 |