Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308020.1 | NP_001294949.1 | 8 - 9 | 890 | 128 |
NM_002647.3 | NP_002638.2 | 9 - 10 | 1079 | 128 | |
XM_011526031.2 | XP_011524333.1 | 10 - 11 | 1244 | 128 | |
GenBank mRNA | AK298517.1 | - | 8 - 9 | 834 | 128 |
AK308364.1 | - | 9 - 10 | 1014 | 128 | |
BC033004.2 | - | 9 - 10 | 1029 | 128 | |
BC053651.1 | - | 9 - 10 | 1009 | 128 |