Assay Design
Gene Transcripts
- Gene Symbol
- ACOT7
- Entrez Gene ID
-
11332
- Gene Name
- acyl-CoA thioesterase 7
- Gene Aliases
-
ACH1, ACT, BACH, CTE-II, LACH, LACH1, hBACH
- Chromosome Location
-
Chr.1: 6264272 - 6393766 on Build GRCh38
- Gene Symbol
- LOC344967
- Entrez Gene ID
-
344967
- Gene Name
- acyl-CoA thioesterase 7 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 40042917 - 40057199 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027277.2 |
|
3 - 3 |
920 |
55 |
GenBank mRNA |
AK125299.1 |
- |
3 - 3 |
920 |
55 |
View Full Product Details