Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288962.1 | NP_001275891.1 | 12 - 13 | 1476 | 64 |
NM_001288963.1 | NP_001275892.1 | 11 - 12 | 1308 | 64 | |
NM_004240.3 | NP_004231.1 | 11 - 12 | 1308 | 64 | |
NR_110231.1 | 12 - 13 | 1327 | 64 | ||
XM_006722940.1 | XP_006723003.1 | 12 - 13 | 1476 | 64 | |
GenBank mRNA | AB007845.1 | - | 11 - 12 | 1283 | 64 |
AB072596.1 | - | 12 - 13 | 1302 | 64 | |
AF502289.1 | - | 12 - 13 | 1435 | 64 | |
AJ000414.1 | - | 11 - 12 | 1267 | 64 | |
AK223109.1 | - | 11 - 12 | 1273 | 64 | |
AK313296.1 | - | 11 - 12 | 1290 | 64 | |
AL832186.1 | - | 12 - 13 | 1328 | 64 | |
AY081141.1 | - | 11 - 12 | 1262 | 64 | |
BC013002.2 | - | 11 - 12 | 1253 | 64 | |
BT006698.1 | - | 11 - 12 | 1228 | 64 | |
BT020167.1 | - | 11 - 12 | 1228 | 64 | |
BT020171.1 | - | 11 - 12 | 1228 | 64 | |
CR536513.1 | - | 11 - 12 | 1228 | 64 |