Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288962.1 | NP_001275891.1 | 13 - 14 | 1607 | 70 |
NM_001288963.1 | NP_001275892.1 | 12 - 13 | 1439 | 70 | |
NM_004240.3 | NP_004231.1 | 12 - 13 | 1439 | 70 | |
NR_110231.1 | 13 - 14 | 1458 | 70 | ||
XM_006722940.1 | XP_006723003.1 | 13 - 14 | 1607 | 70 | |
GenBank mRNA | AB007845.1 | - | 12 - 13 | 1414 | 70 |
AB072596.1 | - | 13 - 14 | 1433 | 70 | |
AF380114.1 | - | 12 - 13 | 1359 | 70 | |
AF502289.1 | - | 13 - 14 | 1566 | 70 | |
AJ000414.1 | - | 12 - 13 | 1398 | 70 | |
AK223109.1 | - | 12 - 13 | 1404 | 70 | |
AK313296.1 | - | 12 - 13 | 1421 | 70 | |
AL832186.1 | - | 13 - 14 | 1459 | 70 | |
BC013002.2 | - | 12 - 13 | 1384 | 70 | |
BT006698.1 | - | 12 - 13 | 1359 | 70 | |
BT020167.1 | - | 12 - 13 | 1359 | 70 | |
BT020171.1 | - | 12 - 13 | 1359 | 70 | |
CR536513.1 | - | 12 - 13 | 1359 | 70 | |
L40379.1 | - | 2 - 3 | 69 | 70 |