Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288962.1 | NP_001275891.1 | 8 - 9 | 869 | 62 |
NM_001288963.1 | NP_001275892.1 | 8 - 9 | 869 | 62 | |
NM_004240.3 | NP_004231.1 | 8 - 9 | 869 | 62 | |
NR_110231.1 | 8 - 9 | 869 | 62 | ||
XM_005259683.2 | XP_005259740.1 | 8 - 9 | 869 | 62 | |
XM_006722940.1 | XP_006723003.1 | 8 - 9 | 869 | 62 | |
GenBank mRNA | AB007845.1 | - | 8 - 9 | 844 | 62 |
AB072596.1 | - | 8 - 9 | 844 | 62 | |
AF380114.1 | - | 8 - 9 | 789 | 62 | |
AF502289.1 | - | 8 - 9 | 828 | 62 | |
AJ000414.1 | - | 8 - 9 | 828 | 62 | |
AK223109.1 | - | 8 - 9 | 834 | 62 | |
AK313296.1 | - | 8 - 9 | 851 | 62 | |
AY081141.1 | - | 8 - 9 | 823 | 62 | |
BC013002.2 | - | 8 - 9 | 814 | 62 | |
BT006698.1 | - | 8 - 9 | 789 | 62 | |
BT020167.1 | - | 8 - 9 | 789 | 62 | |
BT020171.1 | - | 8 - 9 | 789 | 62 | |
CR536513.1 | - | 8 - 9 | 789 | 62 |