Assay Design



Gene Transcripts

Gene Symbol
SLC25A23
Entrez Gene ID
79085
Gene Name
solute carrier family 25 member 23
Gene Aliases
APC2, MCSC2, SCaMC-3
Chromosome Location
Chr.19: 6436081 - 6460221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024103.2 NP_077008.2 2 - 3 427 99
XM_011528274.1 XP_011526576.1 2 - 3 878 99
XM_011528275.1 XP_011526577.1 2 - 3 878 99
XM_011528276.1 XP_011526578.1 2 - 3 878 99
XM_011528277.1 XP_011526579.1 2 - 3 878 99
XM_011528278.1 XP_011526580.1 2 - 3 878 99
XM_011528279.1 XP_011526581.1 2 - 3 878 99
XM_011528280.1 XP_011526582.1 2 - 3 878 99
XM_011528281.1 XP_011526583.1 2 - 3 878 99
XM_011528282.1 XP_011526584.1 2 - 3 878 99
XM_011528283.1 XP_011526585.1 2 - 3 878 99
XM_011528284.1 XP_011526586.1 2 - 3 878 99
XM_011528285.1 XP_011526587.1 2 - 3 878 99
XM_017027285.1 XP_016882774.1 2 - 3 878 99
XM_017027286.1 XP_016882775.1 2 - 3 878 99
XM_017027287.1 XP_016882776.1 2 - 3 878 99
XM_017027288.1 XP_016882777.1 2 - 3 878 99
XM_017027289.1 XP_016882778.1 2 - 3 878 99
XM_017027290.1 XP_016882779.1 2 - 3 878 99
XM_017027291.1 XP_016882780.1 2 - 3 878 99
XM_017027292.1 XP_016882781.1 2 - 3 878 99
GenBank mRNA AJ512835.1 - 2 - 3 392 99
AJ619962.1 - 2 - 3 382 99
AJ619988.1 - 2 - 3 392 99
AK054901.1 - 1 - 2 124 99
AK294514.1 - 2 - 3 285 99
AY750170.1 - 2 - 3 392 99
BC001656.1 - 2 - 3 408 99
BG706035.1 - 2 - 3 454 99

View Full Product Details