Assay Design



Gene Transcripts

Gene Symbol
SLC25A23
Entrez Gene ID
79085
Gene Name
solute carrier family 25 member 23
Gene Aliases
APC2, MCSC2, SCaMC-3
Chromosome Location
Chr.19: 6436081 - 6460221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024103.2 NP_077008.2 5 - 6 785 58
XM_011528274.1 XP_011526576.1 6 - 7 1377 58
XM_011528275.1 XP_011526577.1 5 - 6 1236 58
XM_011528276.1 XP_011526578.1 6 - 7 1377 58
XM_011528277.1 XP_011526579.1 6 - 7 1377 58
XM_011528278.1 XP_011526580.1 6 - 7 1377 58
XM_011528279.1 XP_011526581.1 6 - 7 1377 58
XM_011528280.1 XP_011526582.1 6 - 7 1377 58
XM_011528281.1 XP_011526583.1 6 - 7 1377 58
XM_011528282.1 XP_011526584.1 6 - 7 1377 58
XM_011528283.1 XP_011526585.1 6 - 7 1377 58
XM_011528284.1 XP_011526586.1 6 - 7 1377 58
XM_011528285.1 XP_011526587.1 6 - 7 1377 58
XM_017027285.1 XP_016882774.1 6 - 7 1377 58
XM_017027286.1 XP_016882775.1 5 - 6 1236 58
XM_017027287.1 XP_016882776.1 5 - 6 1236 58
XM_017027288.1 XP_016882777.1 5 - 6 1236 58
XM_017027289.1 XP_016882778.1 6 - 7 1377 58
XM_017027290.1 XP_016882779.1 5 - 6 1236 58
XM_017027291.1 XP_016882780.1 5 - 6 1236 58
XM_017027292.1 XP_016882781.1 5 - 6 1236 58
GenBank mRNA AJ512835.1 - 5 - 6 750 58
AJ619962.1 - 5 - 6 740 58
AJ619988.1 - 5 - 6 750 58
AK054901.1 - 4 - 5 482 58
AK294514.1 - 5 - 6 643 58
AK295484.1 - 1 - 2 113 58
AY750170.1 - 5 - 6 750 58
BC001656.1 - 6 - 7 907 58

View Full Product Details