Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004786.2 | NP_004777.1 | 3 - 4 | 620 | 114 |
NR_024546.1 | 4 - 5 | 725 | 114 | ||
XM_006722580.3 | XP_006722643.1 | 4 - 5 | 1225 | 114 | |
XM_011526263.2 | XP_011524565.1 | 5 - 6 | 1317 | 114 | |
XM_011526264.2 | XP_011524566.1 | 4 - 5 | 464 | 114 | |
XM_017026093.1 | XP_016881582.1 | 5 - 6 | 1317 | 114 | |
XM_017026094.1 | XP_016881583.1 | 4 - 5 | 464 | 114 | |
GenBank mRNA | AB209263.1 | - | 3 - 4 | 358 | 114 |
AF003938.1 | - | 3 - 4 | 576 | 114 | |
AF051896.1 | - | 3 - 4 | 501 | 114 | |
AF052659.1 | - | 3 - 4 | 520 | 114 | |
AK094926.1 | - | 5 - 6 | 1334 | 114 | |
AK313653.1 | - | 3 - 4 | 531 | 114 | |
BC001156.1 | - | 3 - 4 | 564 | 114 | |
DA248363.1 | - | 4 - 5 | 464 | 114 | |
FJ224353.1 | - | 3 - 4 | 576 | 114 |