Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032878.3 | NP_116267.3 | 1 - 2 | 61 | 69 |
NM_198867.1 | NP_942567.1 | 1 - 2 | 61 | 69 | |
GenBank mRNA | AK310704.1 | - | 1 - 2 | 58 | 69 |
AK314150.1 | - | 1 - 2 | 40 | 69 |