Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297701.1 | NP_001284630.1 | 4 - 5 | 292 | 79 |
NM_032878.3 | NP_116267.3 | 3 - 4 | 213 | 79 | |
NM_198867.1 | NP_942567.1 | 3 - 4 | 213 | 79 | |
XM_005259357.4 | XP_005259414.1 | 3 - 4 | 260 | 79 | |
GenBank mRNA | AB273714.1 | - | 4 - 5 | 292 | 79 |
AK304805.1 | - | 3 - 4 | 187 | 79 | |
AK310130.1 | - | 3 - 4 | 187 | 79 | |
AK310704.1 | - | 3 - 4 | 210 | 79 | |
AK314150.1 | - | 3 - 4 | 192 | 79 | |
AK315894.1 | - | 3 - 4 | 181 | 79 | |
BC007601.1 | - | 3 - 4 | 182 | 79 | |
BC105801.1 | - | 3 - 4 | 185 | 79 | |
BM713594.1 | - | 4 - 5 | 288 | 79 |