Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033044.3 | NP_001028216.1 | 5 - 6 | 1148 | 72 |
NM_001033056.3 | NP_001028228.1 | 5 - 6 | 850 | 72 | |
NM_002065.6 | NP_002056.2 | 6 - 7 | 1504 | 72 | |
XM_006711278.1 | XP_006711341.1 | 5 - 6 | 1211 | 72 | |
GenBank mRNA | AK122784.1 | - | 5 - 6 | 1148 | 72 |
AY486122.1 | - | 4 - 5 | 607 | 72 | |
AY513283.1 | - | 5 - 6 | 662 | 72 | |
AY947343.1 | - | 5 - 6 | 708 | 72 | |
BC010037.2 | - | 5 - 6 | 720 | 72 | |
BC011700.2 | - | 5 - 6 | 716 | 72 | |
BC011852.2 | - | 5 - 6 | 727 | 72 | |
BC018992.1 | - | 6 - 7 | 1067 | 72 | |
BC031964.1 | - | 5 - 6 | 1250 | 72 | |
BC051726.1 | - | 5 - 6 | 1913 | 72 | |
BC127883.1 | - | 4 - 5 | 518 | 72 | |
BX537384.1 | - | 5 - 6 | 732 | 72 | |
S70290.1 | - | 4 - 5 | 607 | 72 | |
X59834.1 | - | 5 - 6 | 716 | 72 | |
Y00387.2 | - | 5 - 6 | 726 | 72 |