Assay Design



Gene Transcripts

Gene Symbol
TACR2
Entrez Gene ID
6865
Gene Name
tachykinin receptor 2
Gene Aliases
NK2R, NKNAR, SKR, TAC2R
Chromosome Location
Chr.10: 69404202 - 69416918 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK096906.1 - 1 - 2 365 77
AK291996.1 - 3 - 4 1340 77
AK300649.1 - 3 - 4 1138 77
AY322545.1 - 3 - 4 745 77

View Full Product Details