Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142800.1 | NP_001136272.1 | 39 - 40 | 8260 | 130 |
NM_001292009.1 | NP_001278938.1 | 39 - 40 | 8260 | 130 | |
GenBank mRNA | FJ416331.1 | - | 39 - 40 | 8268 | 130 |
FM209056.1 | - | 39 - 40 | 8212 | 130 |