Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014905.4 | NP_055720.3 | 17 - 18 | 2113 | 67 |
GenBank mRNA | AB020645.1 | - | 17 - 18 | 2099 | 67 |
AF097493.1 | - | 10 - 11 | 841 | 67 | |
AF223943.1 | - | 17 - 18 | 1868 | 67 | |
AF327434.1 | - | 17 - 18 | 2079 | 67 | |
AK092974.1 | - | 3 - 4 | 1624 | 67 | |
AK289747.1 | - | 18 - 19 | 2089 | 67 | |
AK294927.1 | - | 5 - 6 | 731 | 67 | |
AK298266.1 | - | 6 - 7 | 465 | 67 | |
BC038507.1 | - | 18 - 19 | 2089 | 67 |