Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001502.2 | NP_001001502.1 | 4 - 5 | 611 | 60 |
NM_001318035.1 | NP_001304964.1 | 4 - 5 | 611 | 60 | |
NM_001318037.1 | NP_001304966.1 | 3 - 4 | 454 | 60 | |
NM_003085.4 | NP_003076.1 | 3 - 4 | 454 | 60 | |
XM_006714914.3 | XP_006714977.1 | 3 - 4 | 449 | 60 | |
XM_006714915.3 | XP_006714978.1 | 3 - 4 | 361 | 60 | |
XM_006714916.2 | XP_006714979.1 | 3 - 4 | 419 | 60 | |
XM_011534640.2 | XP_011532942.1 | 4 - 5 | 477 | 60 | |
GenBank mRNA | BC002902.2 | - | 4 - 5 | 414 | 60 |
BM543516.1 | - | 3 - 4 | 454 | 60 | |
BT006627.1 | - | 2 - 3 | 163 | 60 | |
CR457027.1 | - | 2 - 3 | 163 | 60 | |
JF960951.1 | - | 4 - 5 | 606 | 60 | |
JF960953.1 | - | 3 - 4 | 449 | 60 | |
S69965.1 | - | 3 - 4 | 217 | 60 |