Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080419.2 | NP_001073888.2 | 3 - 4 | 538 | 60 |
XM_006722145.3 | XP_006722208.1 | 6 - 7 | 4919 | 60 | |
XM_011525400.1 | XP_011523702.1 | 4 - 5 | 607 | 60 | |
XM_017025247.1 | XP_016880736.1 | 6 - 7 | 4919 | 60 | |
XM_017025248.1 | XP_016880737.1 | 5 - 6 | 4821 | 60 | |
XM_017025249.1 | XP_016880738.1 | 5 - 6 | 964 | 60 | |
XM_017025250.1 | XP_016880739.1 | 4 - 5 | 866 | 60 | |
GenBank mRNA | AB051540.1 | - | 3 - 4 | 521 | 60 |
BC053362.1 | - | 3 - 4 | 521 | 60 |