Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308373.1 | NP_001295302.1 | 8 - 9 | 1285 | 65 |
NM_012181.4 | NP_036313.3 | 8 - 9 | 1288 | 65 | |
XM_011527863.1 | XP_011526165.1 | 8 - 9 | 1346 | 65 | |
XM_011527864.1 | XP_011526166.1 | 7 - 8 | 1196 | 65 | |
GenBank mRNA | AK026952.1 | - | - | 1876 | 65 |
AK300571.1 | - | 8 - 9 | 1259 | 65 | |
AK313363.1 | - | 8 - 9 | 1219 | 65 | |
AY225339.1 | - | 7 - 8 | 1149 | 65 | |
AY278607.1 | - | 8 - 9 | 1212 | 65 | |
BC009966.1 | - | 7 - 8 | 992 | 65 | |
BQ953871.1 | - | 2 - 3 | 196 | 65 | |
BX538124.1 | - | 8 - 9 | 1266 | 65 | |
BX647405.1 | - | 8 - 9 | 1265 | 65 | |
BX647720.1 | - | 8 - 9 | 1223 | 65 | |
GQ372970.1 | - | 5 - 6 | 785 | 65 |