Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001572.3 | NP_001563.2 | 10 - 11 | 1727 | 77 |
NM_004029.2 | NP_004020.1 | 9 - 10 | 1640 | 77 | |
NM_004031.2 | NP_004022.2 | 8 - 9 | 1806 | 77 | |
XM_005252906.3 | XP_005252963.1 | 9 - 10 | 1792 | 77 | |
XM_005252907.3 | XP_005252964.1 | 9 - 10 | 1789 | 77 | |
XM_005252909.3 | XP_005252966.1 | 8 - 9 | 1705 | 77 | |
XM_011520066.2 | XP_011518368.1 | 10 - 11 | 1703 | 77 | |
XM_017017674.1 | XP_016873163.1 | 3 - 4 | 605 | 77 | |
GenBank mRNA | AF076494.1 | - | 8 - 9 | 1731 | 77 |
AK303752.1 | - | 8 - 9 | 1538 | 77 | |
BC136555.1 | - | 8 - 9 | 1442 | 77 | |
KC477210.1 | - | 7 - 8 | 1719 | 77 | |
U53830.1 | - | 9 - 10 | 1649 | 77 | |
U53831.1 | - | 8 - 9 | 1562 | 77 | |
U53832.1 | - | 8 - 9 | 1481 | 77 | |
U73036.1 | - | 10 - 11 | 1666 | 77 |