Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001572.3 | NP_001563.2 | 5 - 6 | 825 | 103 |
NM_004029.2 | NP_004020.1 | 5 - 6 | 825 | 103 | |
NM_004031.2 | NP_004022.2 | 3 - 4 | 904 | 103 | |
XM_005252906.3 | XP_005252963.1 | 4 - 5 | 890 | 103 | |
XM_005252907.3 | XP_005252964.1 | 4 - 5 | 887 | 103 | |
XM_005252909.3 | XP_005252966.1 | 4 - 5 | 890 | 103 | |
XM_011520066.2 | XP_011518368.1 | 5 - 6 | 801 | 103 | |
GenBank mRNA | AF076494.1 | - | 3 - 4 | 829 | 103 |
BC136555.1 | - | 3 - 4 | 540 | 103 | |
KC477210.1 | - | 3 - 4 | 904 | 103 | |
U53830.1 | - | 4 - 5 | 747 | 103 | |
U53831.1 | - | 4 - 5 | 747 | 103 | |
U73036.1 | - | 5 - 6 | 764 | 103 |