Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004029.2 | NP_004020.1 | 6 - 7 | 1047 | 143 |
XM_005252909.3 | XP_005252966.1 | 5 - 6 | 1112 | 143 | |
GenBank mRNA | KC477210.1 | - | 4 - 5 | 1126 | 143 |
U53831.1 | - | 5 - 6 | 969 | 143 |