Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018370.2 | NP_060840.2 | 3 - 4 | 807 | 91 |
XM_005269004.2 | XP_005269061.1 | 3 - 4 | 692 | 91 | |
XM_017019578.1 | XP_016875067.1 | 4 - 5 | 929 | 91 | |
GenBank mRNA | AK002121.1 | - | 1 - 2 | 113 | 91 |
AK094923.1 | - | 3 - 4 | 437 | 91 | |
BC013773.1 | - | 1 - 2 | 105 | 91 | |
BC018435.1 | - | 3 - 4 | 783 | 91 |