Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023915.3 | NP_076404.3 | 1 - 2 | 263 | 136 |
GenBank mRNA | AB031548.1 | - | 1 - 2 | 195 | 136 |
AF237763.1 | - | 1 - 2 | 151 | 136 | |
BC112941.1 | - | 1 - 2 | 172 | 136 | |
BC117565.1 | - | 1 - 2 | 172 | 136 | |
BC132888.1 | - | 1 - 2 | 229 | 136 | |
BC132890.1 | - | 1 - 2 | 229 | 136 | |
BP226924.1 | - | 1 - 2 | 367 | 136 |