Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000887.4 | NP_000878.2 | 19 - 20 | 2489 | 68 |
NM_001286375.1 | NP_001273304.1 | 19 - 20 | 2489 | 68 | |
GenBank mRNA | BC038237.1 | - | 19 - 20 | 2401 | 68 |
M81695.1 | - | 19 - 20 | 2432 | 68 |