Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019893.2 | NP_063946.2 | 10 - 11 | 1326 | 76 |
XM_011539970.2 | XP_011538272.1 | 11 - 12 | 1522 | 76 | |
XM_011539971.2 | XP_011538273.1 | 11 - 12 | 1526 | 76 | |
XM_011539972.2 | XP_011538274.1 | 10 - 11 | 1346 | 76 | |
XM_011539973.2 | XP_011538275.1 | 11 - 12 | 1532 | 76 | |
XM_017016435.1 | XP_016871924.1 | 11 - 12 | 1526 | 76 | |
GenBank mRNA | AF250847.1 | - | 10 - 11 | 1269 | 76 |
AF449759.2 | - | 10 - 11 | 1326 | 76 |